2,4-Dichloro-5-methylphenol ≥97%

Supplier: AOBChem USA

Warning

28484-1G 28484-25G 28484-5G
76886-428EA 76.39 USD
76886-428 76886-430 76886-432
2,4-Dichloro-5-methylphenol ≥97%
2,4-Dichloro-5-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆Cl₂O
MW: 177.028 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11520109
CAS Number: 1124-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR