1-Boc-5-bromo-6-methyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

28249-1G 28249-25G 28249-500MG 28249-5G
76886-398EA 144.29 USD
76886-398 76886-400 76886-402 76886-404
1-Boc-5-bromo-6-methyl-1H-indazole ≥97%
1-Boc-5-bromo-6-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₁₃H₁₅BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD16628189
CAS Number: 1305320-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR