2,4-Bis(trifluoromethyl)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(2,4-Bis(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

28191-25G 28191-5G
76886-388EA 310.85 USD
76886-388 76886-390
2,4-Bis(trifluoromethyl)phenylboronic acid pinacol ester ≥97%
2,4-Bis(trifluoromethyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₅BF₆O₂
MW: 340.700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09953467
CAS Number: 1073353-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR