1-Bromo-2,3,4-trifluoro-5-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

28170-25G 28170-5G
76886-378EA 346.92 USD
76886-378 76886-380
1-Bromo-2,3,4-trifluoro-5-iodobenzene ≥97%
1-Bromo-2,3,4-trifluoro-5-iodobenzene
Formula: C₆HBrF₃I
MW: 336.876 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28977279
CAS Number: 530145-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR