2,3-Dichloro-6-(trifluoromethyl)benzyl alcohol ≥95%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
(2,3-Dichloro-6-(trifluoromethyl)phenyl)methanol
28100-25G
28100-5G
76886-354EA
389.36
USD
76886-354
76886-356
2,3-Dichloro-6-(trifluoromethyl)benzyl alcohol ≥95%
2,3-Dichloro-6-(trifluoromethyl)benzyl alcohol
Precaución: for Research Use Only
Formula:
C₈H₅Cl₂F₃O Storage Temperature: Ambient |
MDL Number:
MFCD06660274 CAS Number: 886501-93-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...