1,3-Dibromo-5-iodo-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

28032-1G 28032-25G 28032-500MG 28032-5G
76886-310EA 162.32 USD
76886-310 76886-312 76886-314 76886-316
1,3-Dibromo-5-iodo-2-methylbenzene ≥97%
1,3-Dibromo-5-iodo-2-methylbenzene
Formula: C₇H₅Br₂I
MW: 375.82 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28142633
CAS Number: 704909-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR