2,4-Dichloro-6-fluorobenzyl alcohol ≥95% research grade

Supplier: AOBChem USA

Warning

27952-1G 27952-25G 27952-500MG 27952-5G
76886-296EA 221.73 USD
76886-296 76886-298 76886-300 76886-302
2,4-Dichloro-6-fluorobenzyl alcohol ≥95% research grade
2,4-Dichloro-6-fluorobenzyl alcohol
Formula: C₇H₅Cl₂FO
Storage Temperature: Refrigerator
MDL Number: MFCD28142626
CAS Number: 1615212-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR