2-(4-Fluorophenyl)-5-methylpyridine ≥97% research grade

Supplier: AOBChem USA

Warning

27779-1G 27779-25G 27779-500MG 27779-5G
76886-266EA 124.13 USD
76886-266 76886-268 76886-270 76886-272
2-(4-Fluorophenyl)-5-methylpyridine ≥97% research grade
2-(4-Fluorophenyl)-5-methylpyridine
Formula: C₁₂H₁₀FN
MW: 187.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09037512
CAS Number: 85237-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR