1-Bromo-2-chloro-4-phenoxybenzene ≥97%

Supplier: AOBChem USA

Warning

27701-1G 27701-25G 27701-500MG 27701-5G
76886-240EA 171.87 USD
76886-240 76886-242 76886-244 76886-246
1-Bromo-2-chloro-4-phenoxybenzene ≥97%
1-Bromo-2-chloro-4-phenoxybenzene
Formula: C₁₂H₈BrClO
Storage Temperature: Ambient
MDL Number: MFCD28142609
CAS Number: 1196395-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR