(6-Bromo-2-methyl-pyridin-3-yl)-methanol ≥97%

Supplier: AOBChem USA

Warning

27583-1G 27583-25G 27583-500MG 27583-5G
76886-226EA 320.4 USD
76886-226 76886-228 76886-230 76886-232
(6-Bromo-2-methyl-pyridin-3-yl)-methanol ≥97%
(6-Bromo-2-methyl-pyridin-3-yl)-methanol
Formula: C₇H₈BrNO
MW: 202.049 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16610243
CAS Number: 1227576-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR