(6-Methylsulfanylpyridin-3-Yl)Methanol ≥97%

Supplier: AOBChem USA

Warning

27274-1G 27274-25G 27274-500MG 27274-5G
76886-188EA 260.99 USD
76886-188 76886-190 76886-192 76886-194
(6-Methylsulfanylpyridin-3-Yl)Methanol ≥97%
(6-Methylsulfanylpyridin-3-Yl)Methanol
Formula: C₇H₉NOS
MW: 155.217 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21089555
CAS Number: 101990-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR