1-Bromo-4-fluoro-2-propoxybenzene ≥95% research grade

Supplier: AOBChem USA

Warning

27166-25G 27166-5G
76886-166EA 375.56 USD
76886-166 76886-168
1-Bromo-4-fluoro-2-propoxybenzene ≥95% research grade
1-Bromo-4-fluoro-2-propoxybenzene
Formula: C₉H₁₀BrFO
MW: 233.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18917738
CAS Number: 473417-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR