(2-Chloro-5-(methoxycarbonyl)pyridin-3-yl)boronic acid ≥95% research grade

Supplier: AOBChem USA

Warning

27103-1G 27103-500MG 27103-5G
76886-142EA 1053.48 USD
76886-142 76886-144 76886-146
(2-Chloro-5-(methoxycarbonyl)pyridin-3-yl)boronic acid ≥95% research grade
(2-Chloro-5-(methoxycarbonyl)pyridin-3-yl)boronic acid
Formula: C₇H₇BClNO₄
MW: 215.399 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20441832
CAS Number: 2096332-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR