6-(1-Pyrrolidinyl)pyridine-3-boronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-(Pyrrolidin-1-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(Pyrrolidino)pyridine-5-boronic acid pinacol ester

27089-25G 27089-5G
76886-130EA 622.75 USD
76886-130 76886-132
6-(1-Pyrrolidinyl)pyridine-3-boronic acid pinacol ester ≥97% research grade
6-(1-Pyrrolidinyl)pyridine-3-boronic acid pinacol ester
Formula: C₁₅H₂₃BN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06798273
CAS Number: 933986-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR