2,4,5-Tribromothiazole ≥97%

Supplier: AOBChem USA

Warning

26868-1G 26868-25G 26868-500MG 26868-5G
76886-098EA 224.92 USD
76886-098 76886-100 76886-102 76886-104
2,4,5-Tribromothiazole ≥97%
2,4,5-Tribromothiazole
Formula: C₃Br₃NS
MW: 321.81 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19543764
CAS Number: 57314-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR