[3-(pyridin-4-yl)phenyl]boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: (3-(Pyridin-4-yl)phenyl)boronic acid

26793-1G 26793-25G 26793-500MG 26793-5G
76886-072EA 195.21 USD
76886-072 76886-074 76886-076 76886-078
[3-(pyridin-4-yl)phenyl]boronic acid ≥97% research grade
[3-(pyridin-4-yl)phenyl]boronic acid
Formula: C₁₁H₁₀BNO₂
MW: 199.014 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18418032
CAS Number: 337536-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR