[4-(2-methylbutan-2-yl)phenyl]boronic acid ≥97%

Supplier: AOBChem USA

Warning

26723-1G 26723-500MG 26723-5G
76886-062EA 604.72 USD
76886-062 76886-064 76886-066
[4-(2-methylbutan-2-yl)phenyl]boronic acid ≥97%
[4-(2-methylbutan-2-yl)phenyl]boronic acid
Formula: C₁₁H₁₇BO₂
MW: 192.062 g/mol
Storage Temperature: Ambient
CAS Number: 851199-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR