(2-Bromo-3-chloropyridin-4-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

26656-1G 26656-25G 26656-500MG 26656-5G
76886-050EA 381.93 USD
76886-050 76886-052 76886-054 76886-056
(2-Bromo-3-chloropyridin-4-yl)methanol ≥97%
(2-Bromo-3-chloropyridin-4-yl)methanol
Formula: C₆H₅BrClNO
Storage Temperature: Ambient
MDL Number: MFCD16606981
CAS Number: 1227584-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR