[2-Fluoro-6-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol ≥97%

Supplier: AOBChem USA

Warning

26597-1G 26597-25G 26597-500MG 26597-5G
76886-034EA 439.22 USD
76886-034 76886-036 76886-038 76886-040
[2-Fluoro-6-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol ≥97%
[2-Fluoro-6-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanol
Formula: C₁₄H₂₀BFO₄
MW: 282.116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27756670
CAS Number: 1417736-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR