1-(4-Bromo-2-fluorobenzoyl)piperidine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Bromo-2-fluorobenzoyl)piperidine

26565-25G 26565-5G
76886-022EA 559.1 USD
76886-022 76886-024
1-(4-Bromo-2-fluorobenzoyl)piperidine ≥97%
1-(4-Bromo-2-fluorobenzoyl)piperidine
Formula: C₁₂H₁₃BrFNO
Storage Temperature: Ambient
MDL Number: MFCD09938596
CAS Number: 1017035-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR