(3-Bromo-2-chloropyridin-4-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

26386-1G 26386-25G 26386-500MG 26386-5G
76885-978EA 153.84 USD
76885-978 76885-980 76885-982 76885-984
(3-Bromo-2-chloropyridin-4-yl)methanol ≥97%
(3-Bromo-2-chloropyridin-4-yl)methanol
Formula: C₆H₅BrClNO
Storage Temperature: Ambient
MDL Number: MFCD16606986
CAS Number: 1227502-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR