2-Bromo-4-methoxybenzyl Alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Bromo-4-methoxy-phenyl)methanol

26266-1G 26266-25G 26266-5G
76885-952EA 75.33 USD
76885-952 76885-954 76885-956
2-Bromo-4-methoxybenzyl Alcohol ≥97%
2-Bromo-4-methoxybenzyl Alcohol

Precaución: for Research Use Only

Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11042785
CAS Number: 163190-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR