2-(Benzyloxy)-4-bromoanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(Benzyloxy)-4-bromo-1-methoxybenzene

26054-1G 26054-25G 26054-5G
76885-900EA 88.06 USD
76885-900 76885-902 76885-904
2-(Benzyloxy)-4-bromoanisole ≥97%
2-(Benzyloxy)-4-bromoanisole

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO₂
MW: 293.1558 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09836381
CAS Number: 78504-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR