1-Bromo-2-fluoro-4-iodo-5-nitrobenzene ≥97%

Supplier: AOBChem USA

Warning

26021-1G 26021-25G 26021-5G
76885-874EA 62.6 USD
76885-874 76885-876 76885-878
1-Bromo-2-fluoro-4-iodo-5-nitrobenzene ≥97%
1-Bromo-2-fluoro-4-iodo-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrFINO₂
MW: 345.89 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16295088
CAS Number: 1226808-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR