1-(2,6-Dichloropyridin-3-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Acetyl-2,6-dichloropyridine

25996-1G 25996-25G 25996-5G
76885-844EA 114.58 USD
76885-844 76885-846 76885-848
1-(2,6-Dichloropyridin-3-yl)ethanone ≥97%
1-(2,6-Dichloropyridin-3-yl)ethanone
Formula: C₇H₅Cl₂NO
MW: 190.0267 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11847148
CAS Number: 412018-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR