4-Bromo-2,3-difluoroiodobenzene ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-2,3-difluoro-4-iodobenzene

25944-1G 25944-25G 25944-500MG 25944-5G
76885-828EA 165.51 USD
76885-828 76885-830 76885-832 76885-834
4-Bromo-2,3-difluoroiodobenzene ≥95%
4-Bromo-2,3-difluoroiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrF₂I
MW: 318.885 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878200
CAS Number: 207989-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR