6-Bromo-3-chloro-2-fluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (6-Bromo-3-chloro-2-fluorophenyl)methanol

25940-1G 25940-25G 25940-500MG 25940-5G
76885-812EA 260.99 USD
76885-812 76885-814 76885-816 76885-818
6-Bromo-3-chloro-2-fluorobenzyl alcohol ≥97%
6-Bromo-3-chloro-2-fluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅BrClFO
MW: 239.469 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24848703
CAS Number: 1449008-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR