(4-Fluoro-1H-indazol-6-yl)boronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

25684-1G 25684-500MG 25684-5G 25684-25G
76885-756EA 462.56 USD
76885-756 76885-758 76885-760 77635-562
(4-Fluoro-1H-indazol-6-yl)boronic acid pinacol ester ≥97% research grade
(4-Fluoro-1H-indazol-6-yl)boronic acid pinacol ester
Formula: C₁₃H₁₆BFN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD27756655
CAS Number: 2121511-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR