(6-Ethoxypyridin-3-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

25661-1G 25661-25G 25661-500MG 25661-5G
76885-744EA 221.73 USD
76885-744 76885-746 76885-748 76885-750
(6-Ethoxypyridin-3-yl)methanol ≥97%
(6-Ethoxypyridin-3-yl)methanol
Formula: C₈H₁₁NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD14606407
CAS Number: 101990-67-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR