2-(Boc-amino)-4-chloropyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Chloropyridin-2-yl)carbamic acid tert-butyl ester, tert-Butyl (4-chloropyridin-2-yl)carbamate

25527-25G 25527-5G
76885-720EA 350.1 USD
76885-720 76885-722
2-(Boc-amino)-4-chloropyridine ≥97%
2-(Boc-amino)-4-chloropyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₃ClN₂O₂
MW: 228.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663519
CAS Number: 130721-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR