4'-Methoxy-3'-methylacetophenone ≥97%

Supplier: AOBChem USA

Warning

25480-25G 25480-5G
76885-710EA 396.78 USD
76885-710 76885-712
4'-Methoxy-3'-methylacetophenone ≥97%
4'-Methoxy-3'-methylacetophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD01673307
CAS Number: 10024-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR