4-(3-Bromo-2-fluorobenzyl)morpholine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Morpholine, 4-[(3-bromo-2-fluorophenyl)methyl]-, 1-BROMO-2-FLUORO-3-(MORPHOLINOMETHYL)BENZENE

25428-1G 25428-25G 25428-5G 25428-500MG
76885-700EA 99.73 USD
76885-700 76885-702 76885-704 77649-542
4-(3-Bromo-2-fluorobenzyl)morpholine ≥97%
4-(3-Bromo-2-fluorobenzyl)morpholine

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrFNO
MW: 274.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21333073
CAS Number: 1355247-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR