6-Bromo-2,3-difluorobenzyl alcohol ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
(6-Bromo-2,3-difluorophenyl)methanol
25410-1G
25410-25G
25410-5G
76885-686EA
64.72
USD
76885-686
76885-688
76885-690
6-Bromo-2,3-difluorobenzyl alcohol ≥97%
6-Bromo-2,3-difluorobenzyl alcohol
Precaución: for Research Use Only
Formula:
C₇H₅BrF₂O MW: 223.015 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD07368109 CAS Number: 651326-72-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...