1-(2-Fluoropyridin-3-yl)ethanol ≥95%

Supplier: AOBChem USA

Warning

25402-1G 25402-25G 25402-500MG 25402-5G
76885-670EA 263.11 USD
76885-670 76885-672 76885-674 76885-676
1-(2-Fluoropyridin-3-yl)ethanol ≥95%
1-(2-Fluoropyridin-3-yl)ethanol
Formula: C₇H₈FNO
Storage Temperature: Refrigerator
MDL Number: MFCD16037359
CAS Number: 79574-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR