1-(2-Fluoro-4-iodo-3-pyridinyl)-ethanone ≥97%

Supplier: AOBChem USA

Warning

25359-1G 25359-500MG 25359-5G
76885-648EA 736.27 USD
76885-648 76885-650 76885-652
1-(2-Fluoro-4-iodo-3-pyridinyl)-ethanone ≥97%
1-(2-Fluoro-4-iodo-3-pyridinyl)-ethanone
Formula: C₇H₅FINO
MW: 265.024 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22627837
CAS Number: 1160502-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR