3-Chloro-2,6-difluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Chloro-2,6-difluorophenyl)methanol

25326-25G 25326-5G
76885-630EA 177.18 USD
76885-630 76885-632
3-Chloro-2,6-difluorobenzyl alcohol ≥97%
3-Chloro-2,6-difluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD01631325
CAS Number: 252004-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR