1-Bromo-4-chloro-3-fluoro-2-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

25171-1G 25171-25G 25171-500MG 25171-5G
76885-606EA 231.28 USD
76885-606 76885-608 76885-610 76885-612
1-Bromo-4-chloro-3-fluoro-2-methoxybenzene ≥97%
1-Bromo-4-chloro-3-fluoro-2-methoxybenzene
Formula: C₇H₅BrClFO
MW: 239.46 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22414766
CAS Number: 1414870-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR