2,3-Dimethyl-4-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

25155-1G 25155-25G 25155-500MG 25155-5G
76885-590EA 178.24 USD
76885-590 76885-592 76885-594 76885-596
2,3-Dimethyl-4-methoxyphenylboronic acid ≥97%
2,3-Dimethyl-4-methoxyphenylboronic acid
Formula: C₉H₁₃BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD17926494
CAS Number: 149507-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR