1-(3-Bromo-4-methylbenzyl)pyrrolidine ≥97% research grade

Supplier: AOBChem USA

Warning

24952-1G 24952-25G 24952-500MG 24952-5G
76885-576EA 290.69 USD
76885-576 76885-578 76885-580 76885-582
1-(3-Bromo-4-methylbenzyl)pyrrolidine ≥97% research grade
1-(3-Bromo-4-methylbenzyl)pyrrolidine
Formula: C₁₂H₁₆BrN
Storage Temperature: Ambient
MDL Number: MFCD22628242
CAS Number: 1414870-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR