(3-Ethylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

24718-1G 24718-25G 24718-500MG 24718-5G
76885-522EA 213.25 USD
76885-522 76885-524 76885-526 76885-528
(3-Ethylphenyl)methanol ≥97%
(3-Ethylphenyl)methanol
Formula: C₉H₁₂O
MW: 136.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09999852
CAS Number: 82657-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR