1-Cyclopropyl-2,4-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

24496-1G 24496-500MG 24496-5G
76885-476EA 498.63 USD
76885-476 76885-478 76885-480
1-Cyclopropyl-2,4-difluorobenzene ≥97%
1-Cyclopropyl-2,4-difluorobenzene
Formula: C₉H₈F₂
Storage Temperature: Refrigerator
MDL Number: MFCD22543711
CAS Number: 1394291-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR