1-Bromo-2-iodo-3,5-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

24396-1G 24396-25G 24396-5G
76885-438EA 62.6 USD
76885-438 76885-440 76885-442
1-Bromo-2-iodo-3,5-dimethylbenzene ≥97%
1-Bromo-2-iodo-3,5-dimethylbenzene
Formula: C₈H₈BrI
Storage Temperature: Refrigerator
MDL Number: MFCD07780646
CAS Number: 933671-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR