(5-Methoxypyridin-3-yl)methanol ≥97%
Supplier: AOBChem USA
|
Warning
|
24332-1G
24332-25G
24332-5G
24332-500MG
76885-414EA
91.24
USD
76885-414
76885-416
76885-418
77649-446
(5-Methoxypyridin-3-yl)methanol ≥97%
(5-Methoxypyridin-3-yl)methanol
Formula:
C₇H₉NO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD11052844 CAS Number: 937202-11-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...