2-(2-Methoxy-ethoxy)-5-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(2-Methoxyethoxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(2-Methoxyethoxy)-5-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)pyridine

24211-1G 24211-25G 24211-5G
76885-400EA 79.57 USD
76885-400 76885-402 76885-404
2-(2-Methoxy-ethoxy)-5-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyridine ≥97%
2-(2-Methoxy-ethoxy)-5-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyridine

Precaución: for Research Use Only

Formula: C₁₄H₂₂BNO₄
MW: 279.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16995934
CAS Number: 1257553-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR