(6-Methoxy-3-pyridyl)methanol ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 5-(Hydroxymethyl)-2-methoxypyridine, (6-Methoxypyridin-3-yl)methanol

24142-25G 24142-5G
76885-392EA 289.63 USD
76885-392 76885-394
(6-Methoxy-3-pyridyl)methanol ≥95%
(6-Methoxy-3-pyridyl)methanol
Formula: C₇H₉NO₂
MW: 139.16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06202826
CAS Number: 58584-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR