2-(Methylthio)thiophene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(Mehtylthio)thiophene

23952-25G 23952-5G
76885-382EA 339.49 USD
76885-382 76885-384
2-(Methylthio)thiophene ≥97%
2-(Methylthio)thiophene

Precaución: for Research Use Only

Formula: C₅H₆S₂
MW: 130.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00052382
CAS Number: 5780-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR