2-(4-Pyridyl)benzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2-(4-Pyridinyl)benzaldehyde, 2-(Pyridin-4-yl)benzaldehyde
23923-1G
23923-25G
23923-5G
76885-368EA
62.6
USD
76885-368
76885-370
76885-372
2-(4-Pyridyl)benzaldehyde ≥97%
2-(4-Pyridyl)benzaldehyde
Precaución: for Research Use Only
Formula:
C₁₂H₉NO Storage Temperature: Refrigerator |
MDL Number:
MFCD02684098 CAS Number: 176526-00-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...