1,2-Difluoro-4-(Methylsulfanyl)Benzene ≥97%

Supplier: AOBChem USA

Warning

23917-1G 23917-25G 23917-5G
76885-358EA 66.84 USD
76885-358 76885-360 76885-362
1,2-Difluoro-4-(Methylsulfanyl)Benzene ≥97%
1,2-Difluoro-4-(Methylsulfanyl)Benzene
Formula: C₇H₆F₂S
MW: 160.184 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08669333
CAS Number: 130922-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR