6-(Dimethylamino)nicotinaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-(N,N-Dimethylamino)pyridine-5-carboxaldehyde

23808-1G 23808-25G 23808-500MG 23808-5G
76885-330EA 147.47 USD
76885-330 76885-332 76885-334 76885-336
6-(Dimethylamino)nicotinaldehyde ≥97% research grade
6-(Dimethylamino)nicotinaldehyde
Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08272100
CAS Number: 149805-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR