2,4-Dibromo-6-fluoro-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

23805-25G 23805-5G
76885-318EA 198.39 USD
76885-318 76885-320
2,4-Dibromo-6-fluoro-iodobenzene ≥97%
2,4-Dibromo-6-fluoro-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂FI
Storage Temperature: Ambient
MDL Number: MFCD00143072
CAS Number: 202982-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR